Learn More
Miele Professional™ Miele Process Documentation
Shop All Miele Products
Click to view available options
:
Software; to connect Segosoft
Description
- For the quick and simple assignment of cycles to a patient
- Service Card guarantees compliance with medical devices directive
- The scope of services includes a licence card
- For the paperless transfer of data
Specifications
Specifications
| Description | SegoAssign Plus |
| Product Type | Process/Operating Documentation/Communication |
| For Use With (Equipment) | Washer-disinfectors, laboratory |
| Depth (Metric) | 55 mm |
| Height (Metric) | 1 mm |
| Width (Metric) | 86 mm |