missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIER2 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98516B
This item is not returnable.
View return policy
Description
MIER2 Polyclonal antibody specifically detects MIER2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MIER2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 54531 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Biotin | |
| KIAA1193mesoderm induction early response protein 2, mesoderm induction early response 1, family member 2, Mi-er2 | |
| Produced in rabbits immunized with E. coli-derived Human MIER2 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction