missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIER2 Antibody (OTI3G7), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72683B
This item is not returnable.
View return policy
Description
MIER2 Monoclonal specifically detects MIER2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MIER2 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| MIER2 | |
| Recombinant protein fragment corresponding to amino acids 1-296 of human MIER2 (NP_060020) produced in E.coli. | |
| 0.1 mL | |
| 54531.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI3G7 | |
| Western Blot, Immunohistochemistry | |
| KIAA1193mesoderm induction early response protein 2, mesoderm induction early response 1, family member 2, Mi-er2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur