missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MIER2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550451
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total MIER2 protein.
Transcriptional repressor.
Specifications
| MIER2 | |
| Polyclonal | |
| Unconjugated | |
| Mier2 | |
| 2700087H15Rik; KIAA1193; mesoderm induction early response 1, family member 2; mesoderm induction early response protein 2; MIER family member 2; MIER2; Mi-er2; mKIAA1193; RGD1307278 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54531 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2.2 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q8N344 | |
| Mier2 | |
| Fusion protein corresponding to a region derived from internal residues of human mesoderm induction early response 1, family member 2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction