Learn More
MIF Rabbit, anti-Mouse, Polyclonal, Proteintech
Product Code. 18036175 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18036175 20 μL 20µL
18044745 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18036175 Supplier Proteintech Supplier No. 309571AP20UL
Rabbit Polyclonal Antibody
The MIF antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse MIF. This antibody recognizes mouse, rat antigen. The MIF antibody has been validated for the following applications: FC, IF, WB, ELISA analysis.
Specifications
Antigen MIF
Applications Western Blot, Immunofluorescence, Flow Cytometry, ELISA
Concentration 500 μg/mL
Gene Symbols GIF, Glif, Mif
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 17319
Target Species Mouse, Rat
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG