missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIF4GD Antibody (OTI4C11), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72685UV
This item is not returnable.
View return policy
Description
MIF4GD Monoclonal specifically detects MIF4GD in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| MIF4GD | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MIF4GD | |
| Full length recombinant protein of human MIF4GD (NP_065730) produced in HEK293T cells. | |
| 0.1 mL | |
| 57409.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| OTI4C11 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| AD023, hSLIP1, MGC45027, MIF4G domain containing, MIFD, SLBP (stem loop binding protein)-interacting protein 1, SLBP-interacting protein 1, SLIP1MIF4G domain-containing protein | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction