missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIG (CXCL9) Mouse anti-Human, PE, Clone: B8-11, BD Pharmingen™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566013
This item is not returnable.
View return policy
Description
MIG (CXCL9) Monoclonal antibody specifically detects MIG (CXCL9) in Human samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifications
| MIG (CXCL9) | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 50 Tests | |
| Cell Biology | |
| Human | |
| Purified |
| Flow Cytometry, Flow Cytometry (Intracellular Staining) | |
| B8-11 | |
| Vol. Per test: 5μL | |
| C-X-C motif chemokine 9; CMK; crg-10; Humig; SCYB9 | |
| Recombinant Human MIG Protein | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions and unconjugated antibody and free PE were removed. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction