missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIG2/Kindlin-2 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-36541AF594
This item is not returnable.
View return policy
Description
MIG2/Kindlin-2 Polyclonal antibody specifically detects MIG2/Kindlin-2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Western Blot | |
| fermitin family homolog 2, fermitin family homolog 2 (Drosophila), fermitin family member 2, KIND2DKFZp686G11125, kindlin 2, Kindlin-2, mig-2, MIG2FLJ34213, mitogen inducible gene 2 protein, Mitogen-inducible gene 2 protein, PH domain-containing family C member 1, pleckstrin homology domain containing, family C (with FERM domain) member 1, pleckstrin homology domain containing, family C member 1, Pleckstrin homology domain-containing family C member 1, PLEKHC1FLJ44462, UNC112, UNC112B | |
| RUO | |
| Purified |
| 50mM Sodium Borate | |
| Immunogen affinity purified | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction