Learn More
Migalastat hydrochloride, MedChemExpress
Product Code. 16486596
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486596 100 mg 100mg
16468847 5 mg 5mg
16466258 10 mg 10mg
16422387 50 mg 50mg
16403456 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16486596 Supplier Medchem Express Supplier No. HY14929A
MedChemExpress Migalastat hydrochloride (GR181413A) is a potent and competitive inhibitor of α-galactosidase A (α-Gal A) with an IC50 of 0.04 μM for human α-Gal A.
Migalastat hydrochloride (GR181413A) is a potent and competitive inhibitor of α-galactosidase A (α-Gal A) with an IC50 of 0.04 μM for human α-Gal A.
Specifications
Chemical Name or Material Migalastat hydrochloride
Physical Form Solid
CAS 75172-81-5
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C6H14ClNO4
Synonym GR181413A
Solubility Information H2O : ≥ 200 mg/mL (1001.85 mM)
SMILES O[C@H]1[C@@H](CO)NC[C@H](O)[C@H]1O.Cl
Molecular Weight (g/mol) 199.63
Formula Weight 199.63
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only