missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Migfilin Antibody (FBLIM1/8189R), DyLight 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21075UV
This item is not returnable.
View return policy
Description
Migfilin Monoclonal antibody specifically detects Migfilin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Migfilin | |
| Monoclonal | |
| DyLight 350 | |
| CAL, DKFZp434G171, FBLP-1FBLP1CSX-associated LIM, filamin binding LIM protein 1, filamin-binding LIM protein 1, filamin-binding LIM protein-1, MIG2-interacting protein, Migfilin, mitogen-inducible 2 interacting protein, Mitogen-inducible 2-interacting protein | |
| Recombinant full-length human Migfilin protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| FBLIM1/8189R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 54751 | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction