missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Migfilin Antibody (OTI3G1) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72691
This item is not returnable.
View return policy
Description
Migfilin Monoclonal specifically detects Migfilin in Human samples. It is validated for Western Blot.
Specifications
| Migfilin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000 | |
| CAL, DKFZp434G171, FBLP-1FBLP1CSX-associated LIM, filamin binding LIM protein 1, filamin-binding LIM protein 1, filamin-binding LIM protein-1, MIG2-interacting protein, Migfilin, mitogen-inducible 2 interacting protein, Mitogen-inducible 2-interacting protein | |
| Mouse | |
| 40.5 kDa | |
| 100 μg | |
| 54751 | |
| Human | |
| Lyophilized |
| Western Blot | |
| OTI3G1 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| FBLIM1 | |
| Human recombinant protein fragment corresponding to amino acids 1-242 of human FBLIM1(NP_060026) produced in E.coli. | |
| Affinity Purified | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction