Description
- Dermatologically tested and fresh-scented foam hand soap.
- A gentle soap that helps prevent dry skin with moisturising, lipid-replenishing ingredients and a pearly shine.
- 96% of ingredients are from natural origin.
- Minimise energy use - this soap is proven effective in cold water (based on testing at 20 degrees Celcius).
Specifications
Specifications
| Product Type | Hand Cleanser |
| Color | Light Yellow |
| Form | Liquid |
| Packaging | Plastic Bottle |
| Quantity | 8/Pk. |
| Size | 525 mL |