Learn More
Millesimal Calibration Buffer pH 7.01, Hanna Instruments™
Specifications
Specifications
| Color | Colorless |
| pH | 7.01 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Solution Type | Buffer |
| Formulation | Aqueous Buffer Solution |
| Odor | Odorless |