missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mimecan Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112635
This item is not returnable.
View return policy
Description
Mimecan Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot
Mimecan induces ectopic bone formation in conjunction with transforming growth factor beta. Mimecan is a small proteoglycan which contains tandem leucine-rich repeats. Mimecan is encoded by the OGN gene, a member of the small leucine-rich proteoglycan (SLRP) family of proteins. Mimecan induces ectopic bone formation in conjunction with transforming growth factor beta (TGF-beta-1 or TGF-beta-2) and may regulate osteoblast differentiation. High expression of mimecan protein may be associated with elevated heart left ventricular mass. Alternative splicing results in multiple transcript variants of mimecan. Alternatively spliced transcript variants that encode mimecan have been found.
Specifications
| Mimecan | |
| Polyclonal | |
| Unconjugated | |
| OGN | |
| 3110079A16Rik; corneal keratan sulfate proteoglycan; mimecan; mimecan proteoglycan; mimican; OG; Ogn; OIF; Osteoglycin; osteoglycin/mimecan; osteoinductive factor; SLRR3A | |
| Rabbit | |
| Protein G | |
| RUO | |
| 291015, 4969 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P20774 | |
| OGN | |
| Recombinant Human Mimecan protein (21-298AA). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction