missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MINA Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21502H
This item is not returnable.
View return policy
Description
MINA Polyclonal antibody specifically detects MINA in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MINA | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| DKFZp762O1912, FLJ14393, MDIGDKFZp762O1912, MINA53mdig, mineral dust induced gene protein, Mineral dust-induced gene protein, MYC induced nuclear antigen, MYC-induced nuclear antigen, myc-induced nuclear antigen, 53 kDa, NO52, Nucleolar protein 52 | |
| Myosin Heavy Chain 3 antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human Myosin MINA. The immunogen is located within amino acids 380 - 430 of MINA. Amino Acid Squence: HSLKNSRETHMMGNE | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 84864 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction