missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MINDY3 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2504UV
This item is not returnable.
View return policy
Description
FAM188A Polyclonal antibody specifically detects FAM188A in Human samples. It is validated for ELISA
Specifications
| FAM188A | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 80013 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| DyLight 350 | |
| C10orf97caspase recruitment domain containing pro-apoptotic protein, CARPRP11-394I23.1, chromosome 10 open reading frame 97, Dermal papilla-derived protein 5, DERP5CARD-containing protein, family with sequence similarity 188, member A, FLJ13397, MST126, my042, Protein CARP | |
| Residues 59-76 AFLLKKLLFSSEKSSWRD of human CARP protein. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction