Learn More
Mineral Oil, Contains Vitamin E as a stabilizer in the range of 10 to 100ppm, U.S.P., J.T. Baker™
Product Code. 15568044 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15568044 500 mL Narrow Mouth Amber Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15568044 Supplier Avantor J.T.Baker Supplier No. 270501
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Specifications
CAS 8012-95-1
Flash Point 135.00°C
Packaging Narrow Mouth Amber Glass Bottle
Molecular Formula MFCD00131611
MDL Number MFCD00131611
Quantity 500 mL
Specific Gravity 1L = 0.85kg
Synonym delphinidin chloride, delphinidin, delphinidine, delphinidol, ephdine, delfinidol chloride, unii-em6md4aehe, ccris 2518, em6md4aehe, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride
InChI Key FFNDMZIBVDSQFI-UHFFFAOYSA-N
SMILES *
IUPAC Name 2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride
Molecular Weight (g/mol) 0.00
PubChem CID 68245
ChEBI CHEBI:38701
Grade USP
Chemical Name or Material Mineral Oil, Contains Vitamin E as a stabilizer in the range of 10 to 100ppm
Show More Show Less
compliance-icons
Product Identifier
  • Mineral Oil
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Paraffin oils