Learn More
Mineral Oil NF White, Macron Fine Chemicals™
Product Code. 15548374
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Glass
Unit Size:
4 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15548374 4 L Glass 4 L
15538374 500 mL Glass 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15548374 Supplier Avantor Macron Supplier No. 635810
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Chemical Identifiers
CAS 8012-95-1
Molecular Formula MFCD00131611
Molecular Weight (g/mol) 0.00
MDL Number MFCD00131611
InChI Key FFNDMZIBVDSQFI-UHFFFAOYSA-N
Synonym delphinidin chloride, delphinidin, delphinidine, delphinidol, ephdine, delfinidol chloride, unii-em6md4aehe, ccris 2518, em6md4aehe, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride
PubChem CID 68245
ChEBI CHEBI:38701
IUPAC Name 2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride
SMILES *
Specifications
Color White
Packaging Glass
Quantity 4 L
Grade NF
Physical Form Liquid
Chemical Name or Material Mineral Oil, Light
compliance-icons
Product Identifier
  • Mineral Oil
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Paraffin oils