Learn More
B Braun™ Mini Spike™ V Dispensing Pin
B Braun™ Mini Spike™ Dispensing Pin can be used to withdraw or inject fluids from medication vials or semi-rigid containers.
Brand: B Braun™ 4550560
Additional Details : Weight : 0.00090kg
Description
- Includes 0.45μm bacteria retentive air filter to reduce contamination
- Integrated air channel enables automation of filter pressure balancing technique steps, which singnificantly increases efficiency and safety
- Designed with a snap cap to reduce touch contamination by allowing user to work away from the critical luer connection
- Molded tight-sealing cap improves working hygiene by preventing cap loss and potential leakage when closed
- PVC-, DEHP, and latex-free
- Needle-free solution to withdraw or injection fluids
- High flow rate
- Ergonomic design
- 2 distinct air and fluid channels
- Universal spike for medication vial
- Large grip plate for convenient and intuitive handling
- Integrated two way valve that protects against free-flow when the system is turned upside down
Specifications
Yes | |
Mini Spike Dispensing Pin with Valve |