Learn More
Ekastu Safety™ Eye Wash Bottle with Funnel
Product Code. p-8009196
Click to view available options
Product Type:
Eyewash Bottle
Eyewash Bottle, Empty
Unit Size:
Each
Product Code. 10385722
Brand: Ekastu Safety™ 377951
Made for immediate rinsing of the eyes after accidental splashing from acids, bases and other hazardous chemicals. EKASTU Safety™ Eye Wash Bottle with Funnel can be used in either upright or horizontal positions.
  • Eye Wash Bottle with funnel
  • DIN EN 15154-4
  • Supplied empty or filled
  • For immediate rinsing of the eyes after corrosion by acids, bases and other harsh chemicals
  • PE bottle with screwed-on eye funnel and tilting valve
  • Can be used in both upright and horizontal positions
  • leak-proof
  • Stores easily in EPS Eye Wash Station (sold separately)
Specifications
Capacity (Metric) 600 mL
For Use With (Application) For Immediate Rinsing of the Eyes after Contact with Acids, Bases
Includes Screwed-on Eye Funnel and Tilting Cap
Product Type Eyewash Bottle, Empty