Learn More
Cole-Parmer Miniature Check Valves, Hose Barb, Nylon and Silicone
Product Code. p-200105128 Shop All Cole-Parmer Products
Click to view available options
Inlet:
1/4 in. hose barb
1/8 in. hose barb
3/16 in. hose barb
Unit Size:
Each
Product Code. 30053887
Brand: Cole-Parmer 5011568
Only null left
Add to basket
Add to basket
Versatile check valves can be installed in any position
Prevent backflow to protect your equipment. Work with both liquids and gases. Use for pressure or vacuum applications. 0.18 psi breaking pressure
Specifications
Tubing Size 0.1875 in.
Flow Capacity 0.13
Inlet 3/16 in. hose barb
Outlet Port Barbed
Material (Seal) Silicone