Learn More
Cytiva Minimate™ Tangential Flow Filtration (TFF) Capsule with Omega™ Membrane
Click to view available options
MWCO:
1 kDa
10 kDa
100 kDa
1000 kDa
3 kDa
30 kDa
300 kDa
5 kDa
50 kDa
500 kDa
70 kDa
Unit Size:
Each
Description
- Efficient processing
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
| Material (Membrane) | PES |
| MWCO | 100 kDa |
| Sterility | Non-sterile |
| Filtration Area | 50 cm2 |
| No. per Pack | 1/Pk. |