Learn More
Gilson™ Minipuls Evolution™ Laboratory Peristaltic Pump
Shop All Gilson Products
Click to view available options
Voltage:
100/240 VAC
Unit Size:
Each
Description
- Safe and durable design
- Stainless-steel rollers for great durability and chemical resistance
- Low operating noise
Specifications
Specifications
| For Use With (Application) | Liquid transfer applications |
| Includes | Drive module, Standard accessory package |
| Model | Speed control module |
| Overall Width (Metric) | 164 mm |
| Overall Height (Metric) | 169 mm |
| Shipping Weight (Metric) | 2.4 kg |
| Electrical Requirements | Line Frequency: 50 to 60 Hz / Line Voltage: 100 to 240 VAC / Power consumption:full load 48 w at 220 V – no load 35 w at 220 V |
| Voltage | 100/240 VAC |
| Frequency | 50/60 Hz |