missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MINOS1 Antibody (OTI4H7), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72692AF750
This item is not returnable.
View return policy
Description
MINOS1 Monoclonal specifically detects MINOS1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MINOS1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| MINOS1 | |
| Full length recombinant protein of human MINOS1 (NP_001007535) produced in HEK293T cells. | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI4H7 | |
| Western Blot, Immunohistochemistry | |
| mitochondrial inner membrane organizing system 1 | |
| Mouse | |
| 0.1 mL | |
| 440574 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction