missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIOX Antibody (OTI4G7), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72697PEATT594
This item is not returnable.
View return policy
Description
MIOX Monoclonal specifically detects MIOX in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| MIOX | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| MIOX | |
| Full length recombinant protein of human MIOX (NP_060054) produced in HEK293T cells. | |
| 0.1 mL | |
| 55586.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI4G7 | |
| Flow Cytometry | |
| aldehyde reductase (aldose reductase) like 6, Aldehyde reductase-like 6, ALDRL6, inositol oxygenase, Kidney-specific protein 32, KSP32, MGC90217, MI oxygenase, myo-inositol oxygenaseEC 1.13.99.1, Renal-specific oxidoreductase, RSOR | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction