missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIOX Monoclonal Antibody (OTI3F11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501422S
This item is not returnable.
View return policy
Description
MIOX Monoclonal antibody specifically detects MIOX in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| MIOX | |
| Monoclonal | |
| 0.48 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UGB7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55586 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunocytochemistry, Western Blot | |
| OTI3F11 | |
| Unconjugated | |
| MIOX | |
| 0610009I10Rik; AI314022; aldehyde reductase (aldose reductase) like 6; aldehyde reductase (aldose reductase)-like 6; aldehyde reductase 6, renal; aldehyde reductase-like 6; Aldrl6; C85427; Inositol oxygenase; kidney-specific protein 32; KSP32; MI oxygenase; Miox; Myo-inositol oxygenase; renal-specific oxido-reducatse; renal-specific oxidoreductase; renal-specific oxido-reductase; Rsor | |
| Full length human recombinant protein of human MIOX produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction