missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MIOX Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113351
This item is not returnable.
View return policy
Description
MIOX Polyclonal Antibody for Western Blot, IP, ELISA
MIOX (Myo-Inositol Oxygenase) is a Protein Coding gene. Among its related pathways are superpathway of inositol phosphate compounds and Porphyrin and chlorophyll metabolism. Gene Ontology (GO) annotations related to this gene include iron ion binding and oxidoreductase activity, acting on NAD(P)H.
Specifications
| MIOX | |
| Polyclonal | |
| Unconjugated | |
| MIOX | |
| 0610009I10Rik; AI314022; aldehyde reductase (aldose reductase) like 6; aldehyde reductase (aldose reductase)-like 6; aldehyde reductase 6, renal; aldehyde reductase-like 6; Aldrl6; C85427; Inositol oxygenase; kidney-specific protein 32; KSP32; MI oxygenase; Miox; Myo-inositol oxygenase; renal-specific oxido-reducatse; renal-specific oxidoreductase; renal-specific oxido-reductase; Rsor | |
| Rabbit | |
| Protein G | |
| RUO | |
| 55586, 56727 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9QXN5, Q9UGB7 | |
| MIOX | |
| Recombinant Human Inositol oxygenase protein (1-285AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction