missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIP-1 alpha/CCL3 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | MIP-1 alpha/CCL3 |
|---|---|
| Concentration | 300 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30233359
|
Proteintech
27441-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236846
|
Proteintech
27441-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MIP-1 alpha/CCL3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MIP-1 alpha/CCL3. This antibody recognizes human antigen. The MIP-1 alpha/CCL3 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| MIP-1 alpha/CCL3 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| G0S19-1; LD78ALPHA; MIP-1-alpha; MIP1A; SCYA3 | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 300 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 6348 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title