missing translation for 'onlineSavingsMsg'
Learn More

PeproTech MIP-1 delta Polyclonal Antibody, PeproTech®, Invitrogen™
GREENER_CHOICE

Product Code. 17864403
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
Unit Size:
100µg
1mg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17864403 1 mg 1mg
17894673 50 μg 50µg
17814493 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17864403 Supplier PeproTech Supplier No. 500P123G1MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

AA Sequence of recombinant protein: QFTNDAETEL MMSKLPLENP VVLNSFHFAA DCCTSYISQS IPCSLMKSYF ETSSECSKPG VIFLTKKGRQ VCAKPSGPGV QDCMKKLKPY SI. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human MIP-5 (CCL15). Anti-Human MIP-5 (CCL15)-specific antibody was purified by affinity chromatography employing an immobilized Human MIP-5 (CCL15) matrix. Sandwich ELISA: To detect Human MIP-5 (CCL15) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human MIP-5 (CCL15) (500-P123GBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human MIP-5 (CCL15). Western Blot: To detect Human MIP-5 (CCL15) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human MIP-5 (CCL15) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

This gene is located in a cluster of similar genes in the same region of chromosome 17. These genes encode CC cytokines, which are secreted proteins characterized by two adjacent cysteines. The product of this gene is chemotactic for T cells and monocytes, and acts through C-C chemokine receptor type 1 (CCR1). The proprotein is further processed into numerous smaller functional peptides. Naturally-occurring readthrough transcripts occur from this gene into the downstream gene, CCL14 (chemokine (C-C motif) ligand 14).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIP-1 delta
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CCL15
Gene Accession No. Q16663
Gene Alias C-C motif chemokine 15; C-C motif chemokine ligand 15; CCL15; CCL15(22-92); CCL15(25-92); CCL15(29-92); chemokine (C-C motif) ligand 15; chemokine CC-2; HCC-2; HCC-2Lkn-1; HMRP-2B; leukotactin 1; Leukotactin-1; LKN1; LKN-1; Macrophage inflammatory protein 5; MIP-1 delta; MIP-1D; MIP5; MIP-5; MIP-5MIP-1d; Mrp-2b; NCC3; NCC-3; NCC-3MIP-1D; NCC3MRP-2B; new CC chemokine 3; SCYA15; SCYA15member 15; SCYL3; small inducible cytokine subfamily A (Cys-Cys), member 15; small-inducible cytokine A15; SY15
Gene Symbols CCL15
Host Species Goat
Immunogen E.coli-derived Recombinant Human MIP-5 (CCL15).
Purification Method Antigen affinity chromatography
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6359
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.