missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIPEP Antibody (OTI5E7), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72706R
This item is not returnable.
View return policy
Description
MIPEP Monoclonal specifically detects MIPEP in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| MIPEP | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MIPEP | |
| Recombinant protein fragment corresponding to amino acids 174-516 of human MIPEP (NP_005923) produced in E.coli. | |
| 0.1 mL | |
| 4285.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI5E7 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| HMIP, MIPEC 3.4.24.59, mitochondrial intermediate peptidase | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction