missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIPEP Antibody (OTI5E7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45861
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
MIPEP Monoclonal specifically detects MIPEP in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence.
Specifications
| MIPEP | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:150, Immunofluorescence 1:100 | |
| HMIP, MIPEC 3.4.24.59, mitochondrial intermediate peptidase | |
| Mouse | |
| 80.4 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
| OTI5E7 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| MIPEP | |
| Human recombinant protein fragment corresponding to amino acids 174-516 of human MIPEP (NP_005923) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| 4285 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction