missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MIPEP Monoclonal Antibody (OTI4D11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526415
This item is not returnable.
View return policy
Description
MIPEP Monoclonal Antibody for Western Blot, IHC (P)
The product of this gene performs the final step in processing a specific class of nuclear-encoded proteins targeted to the mitochondrial matrix or inner membrane. This protein is primarily involved in the maturation of oxidative phosphorylation (OXPHOS)-related proteins. This gene may contribute to the functional effects of frataxin deficiency and the clinical manifestations of Friedreich ataxia.
Specifications
| MIPEP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q01992, Q99797 | |
| MIPEP | |
| Human recombinant protein fragment corresponding to amino acids 174-516 of MIPEP produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4D11 | |
| Unconjugated | |
| MIPEP | |
| HMIP; MIP; MIPEP; Mitochondrial intermediate peptidase | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4285, 81684 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction