Learn More
MIPEP Mouse anti-Human, Clone: OTI3G4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF800278
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The product of this gene performs the final step in processing a specific class of nuclear-encoded proteins targeted to the mitochondrial matrix or inner membrane. This protein is primarily involved in the maturation of oxidative phosphorylation (OXPHOS)-related proteins. This gene may contribute to the functional effects of frataxin deficiency and the clinical manifestations of Friedreich ataxia.Specifications
| MIPEP | |
| Monoclonal | |
| Unconjugated | |
| MIPEP | |
| COXPD31, HMIP, MIP | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4285 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI3G4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q99797 | |
| MIPEP | |
| Human recombit protein fragment corresponding to amino acids 174-516 of human MIPEP produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.