missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIPOL1 Antibody (1B7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00145282-M04
This item is not returnable.
View return policy
Description
MIPOL1 Monoclonal antibody specifically detects MIPOL1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA
Specifications
| MIPOL1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| DKFZp313M2036, MGC34010, mirror-image polydactyly 1, mirror-image polydactyly gene 1 protein | |
| MIPOL1 (NP_620059, 321 a.a. ∽ 423 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ARETAVQQYKKLEEEIQTLRVYYSLHKSLSQEENLKDQFNYTLSTYEEALKNRENIVSITQQQNEELATQLQQALTERANMELQLQHAREASQVANEKVQKLE | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Sandwich ELISA | |
| 1B7 | |
| Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA | |
| NP_620059 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 145282 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction