missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Miransertib, MedChemExpress
Description
Miransertib (ARQ-092) is a potent, orally active, selective and allosteric Akt inhibitor with IC50s of 2.7 nM, 14 nM and 8.1 nM for Akt1, Akt2, Akt3, respectively. Miransertib is also a potent the AKT1-E17K mutant protein inhibitor and has the potential for PI3K/AKT-driven tumors and Proteus syndrome research. Miransertib is effective against Leishmania.
Specifications
Specifications
| Chemical Name or Material | Miransertib |
| CAS | 1313881-70-7 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C27H24N6 |
| Quantity | 50 mg |
| Synonym | ARQ-092 |
| Solubility Information | DMSO : 12.5 mg/mL (28.90 mM; Need ultrasonic) |
| SMILES | NC1=NC=CC=C1C2=NC3=CC=C(C4=CC=CC=C4)N=C3N2C5=CC=C(C6(N)CCC6)C=C5 |
| Molecular Weight (g/mol) | 432.52 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?