missing translation for 'onlineSavingsMsg'
Learn More

Miransertib, MedChemExpress

Product Code. 16477676
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477676 100 mg 100mg
16483807 50 mg 50mg
16461849 1 mg 1mg
16440328 5 mg 5mg
16407868 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16477676 Supplier Medchem Express Supplier No. HY19719

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Miransertib (ARQ-092) is a potent, orally active, selective and allosteric Akt inhibitor with IC50s of 2.7 nM, 14 nM and 8.1 nM for Akt1, Akt2, Akt3, respectively. Miransertib is also a potent the AKT1-E17K mutant protein inhibitor and has the potential for PI3K/AKT-driven tumors and Proteus syndrome research. Miransertib is effective against Leishmania.

Miransertib (ARQ-092) is a potent, orally active, selective and allosteric Akt inhibitor with IC50s of 2.7 nM, 14 nM and 8.1 nM for Akt1, Akt2, Akt3, respectively. Miransertib is also a potent the AKT1-E17K mutant protein inhibitor and has the potential for PI3K/AKT-driven tumors and Proteus syndrome research. Miransertib is effective against Leishmania.

Specifications

Chemical Name or Material Miransertib
CAS 1313881-70-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C27H24N6
Quantity 100 mg
Synonym ARQ-092
Solubility Information DMSO : 12.5 mg/mL (28.90 mM; Need ultrasonic)
SMILES NC1=NC=CC=C1C2=NC3=CC=C(C4=CC=CC=C4)N=C3N2C5=CC=C(C6(N)CCC6)C=C5
Molecular Weight (g/mol) 432.52
Formula Weight 432.52
Percent Purity 96.28%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.