missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIS/AMH Antibody (AMH/6713R), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08459B
This item is not returnable.
View return policy
Description
MIS/AMH Monoclonal antibody specifically detects MIS/AMH in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MIS/AMH | |
| Monoclonal | |
| Biotin | |
| MIF, MIS, Muellerian hormone, muellerian-inhibiting factor, muellerian-inhibiting substance, Mullerian hormone, Mullerian inhibiting factor, Mullerian inhibiting substance | |
| A synthetic peptide from the C-terminus of human MIS/AMH (aa 460-560) (exact sequence is proprietary) (Uniprot: IDP03971) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| AMH/6713R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 268 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction