missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIS12 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2359AF350
This item is not returnable.
View return policy
Description
MIS12 Polyclonal antibody specifically detects MIS12 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| MIS12 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry | |
| 2510025F08Rik, hMis12, homolog of yeast Mis12, KNTC2AP, MGC2488, MIS12 homolog, MIS12, MIND kinetochore complex component, homolog (S. pombe), MIS12, MIND kinetochore complex component, homolog (yeast), MTW1, protein MIS12 homolog | |
| The immunogen recognized by this antibody maps to a region between residue 150 and the C-terminus (residue 205) of human MIS12 homolog using the numbering given in entry NP_076944.1 (GeneID 79003). | |
| 0.1 mL | |
| Cell Cycle and Replication, DNA Repair, modENCODE Antibodies | |
| 79003 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction