Learn More
MIS12 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-2358AF594
Product Code. 18698089
  • € 633.00 / 0.10mL
Description
MIS12 Polyclonal antibody specifically detects MIS12 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MIS12
Polyclonal
50mM Sodium Borate
Western Blot
Western Blot
Store at 4C in the dark.