missing translation for 'onlineSavingsMsg'
Learn More

MIS12 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18636749 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18636749 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18636749 Supplier Novus Biologicals Supplier No. NB1002358AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MIS12 Polyclonal antibody specifically detects MIS12 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIS12
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias 2510025F08Rik, hMis12, homolog of yeast Mis12, KNTC2AP, MGC2488, MIS12 homolog, MIS12, MIND kinetochore complex component, homolog (S. pombe), MIS12, MIND kinetochore complex component, homolog (yeast), MTW1, protein MIS12 homolog
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human MIS12 homolog using the numbering given in entry NP_076944.1 (GeneID 79003).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, DNA Repair, modENCODE Antibodies
Primary or Secondary Primary
Gene ID (Entrez) 79003
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.