missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MISA Standard 3: Sb, Sn, Te @ 100 μg/mL in 20% HCl, VHG™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
MISA Standard 3: Sb, Sn, Te @ 100 μg/mL in 20% HCl
Specifications
Specifications
| Chemical Name or Material | Tellurium/Antimony/Tin |
| CAS | 13494-80-9, 7440-36-0, 7440-31-5 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Tellurium/Antimony/Tin |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HCl |
| Quantity | 500 mL |
| Concentration or Composition (by Analyte or Components) | Tellurium 100 μg/mL/Antimony 100.1 μg/mL/Tin 100.1 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction