missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIST1 Antibody (6E8/A12/C11P1), Novus Biologicals™
Mouse Monoclonal Antibody
€ 200.00
Specifications
| Antigen | MIST1 |
|---|---|
| Clone | 6E8/A12/C11P1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:500, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Description
MIST1 Monoclonal specifically detects MIST1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| MIST1 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| basic helix-loop-helix domain containing, class B, 8, basic helix-loop-helix family, member a15, bHLHa15, bHLHb8, class A basic helix-loop-helix protein 15, Class B basic helix-loop-helix protein 8, class II bHLH protein MIST1, MIST1MIST-1, Muscle, intestine and stomach expression 1 | |
| BHLHA15 | |
| IgG1 | |
| Protein A purified | |
| This antibody reacts with Human and Mouse. |
| 6E8/A12/C11P1 | |
| Western Blot 1:100 - 1:500, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q7RTS1 | |
| 168620 | |
| Mouse Mist1, C-terminus residues 175-197. | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title