missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mist1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583883
This item is not returnable.
View return policy
Description
Immunogen sequence: AKNYIKSLTA TILTMSSSRL PGLEGPGPKL YQHYQQQQQV AGGALGATEA QPQGHLQRYS TQIHSFREG Highest antigen sequence indentity to the following orthologs: Mouse - 80%, Rat - 80%.
Plays a role in controlling the transcriptional activity of MYOD1, ensuring that expanding myoblast populations remain undifferentiated. Repression may occur through muscle-specific E-box occupancy by homodimers. May also negatively regulate bHLH-mediated transcription through an N-terminal repressor domain. Serves as a key regulator of acinar cell function, stability, and identity. Also required for normal organelle localization in exocrine cells and for mitochondrial calcium ion transport. May function as a unique regulator of gene expression in several different embryonic and postnatal cell lineages. Binds to the E-box consensus sequence 5'-CANNTG-3'.
Specifications
| Mist1 | |
| Polyclonal | |
| Unconjugated | |
| Bhlha15 | |
| 1810009C13Rik; basic helix-loop-helix domain containing, class B, 8; basic helix-loop-helix family member a15; basic helix-loop-helix family, member a15; Basic helix-loop-helix transcription factor (bHlH); bHlH; BHLHA15; BHLHB8; Class A basic helix-loop-helix protein 15; class B basic helix-loop-helix protein 8; class II bHLH protein MIST1; MIST1; MIST-1; Muscle, intestine and stomach expression 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 168620 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7RTS1 | |
| Bhlha15 | |
| Recombinant protein corresponding to Human BHLHA15. Recombinant protein control fragment (Product #RP-99943). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction