Learn More
Manutan MIT5-D32.B22 - Manometer, 100 mm, 10 bar
Brand:  Manutan A276820
Encompass_Preferred
MIT5-D32.B22 - Manometer, 100 mm, 10 bar
Product Code. 16877020
  • € 54.45 / Each
Only null left
Estimated Shipment: 14-01-2026