missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MITF Antibody (21D1418), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56561AF647
This item is not returnable.
View return policy
Description
MITF Monoclonal specifically detects MITF in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| MITF | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| BHLHE32, bHLHe32MI, Class E basic helix-loop-helix protein 32, microphthalmia-associated transcription factor, Waardenburg syndrome, type 2A, WS2A | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 4286 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 21D1418 | |
| Western Blot | |
| O75030 | |
| MITF | |
| A synthetic peptide mapping to the 400-450 a.a. region of accession number O75030. | |
| 0.1 mL | |
| Primary | |
| Mitf produces, by alternative splicing, 10 different transcripts, all with introns, putatively encoding 10 different protein isoforms. Therefore, observed molecular weights may vary depending on MITF isoforms expressed. Users are encouraged to utilize the peptide immunogen sequence to gain further information about the isoforms recognized by the antibody. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction