missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MITF Antibody (MITF/915), DyLight 550 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11593R
This item is not returnable.
View return policy
Description
MITF Monoclonal antibody specifically detects MITF in Human, Canine, Mouse (Negative), Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MITF | |
| Monoclonal | |
| DyLight 550 | |
| BHLHE32, bHLHe32MI, Class E basic helix-loop-helix protein 32, microphthalmia-associated transcription factor, Waardenburg syndrome, type 2A, WS2A | |
| Recombinant full-length human MITF protein (Uniprot: O75030) | |
| 0.1 mL | |
| Primary | |
| Human, Canine, Mouse (Negative), Rat (Negative) | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MITF/915 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 4286 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction