missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mitochondria Antibody (MTC02/2860R), DyLight 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08548V
This item is not returnable.
View return policy
Description
Mitochondria Monoclonal antibody specifically detects Mitochondria in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Mitochondria | |
| Monoclonal | |
| DyLight 405 | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| MTC02/2860R | |
| 50mM Sodium Borate | |
| Semi-Purified mitochondrial preparation | |
| 100 μg | |
| Cellular Markers, Mitochondrial Markers | |
| Human, Mouse (Negative), Rat (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction