missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mitochondria Antibody (MTC719), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34687MFV500
This item is not returnable.
View return policy
Description
Mitochondria Monoclonal antibody specifically detects Mitochondria in Human samples. It is validated for Flow Cytometry
Specifications
| Mitochondria | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mitochondrial fraction of HeLa cells | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| MTC719 | |
| Flow Cytometry | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction