missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mitochondria Antibody (MTC754), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34688JF549
This item is not returnable.
View return policy
Description
Mitochondria Monoclonal antibody specifically detects Mitochondria in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Specifications
| Mitochondria | |
| Monoclonal | |
| Janelia Fluor 549 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| MTC754 | |
| 50mM Sodium Borate | |
| Mitochondrial fraction of HeLa cells | |
| 0.1 mL | |
| Cellular Markers, Mitochondrial Markers | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction