Learn More
Mitochondria Antibody (MTC754), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-32874-0.02mg
 View more versions of this product
Product Code. 18478981
  • € 290.00 / 0.02mg
Description
Mitochondria Monoclonal antibody specifically detects Mitochondria in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mitochondria
Monoclonal
0.2 mg/mL
Western Blot 0.25-0.5 ug/mL, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL
Mouse
Protein A or G purified
RUO
Primary
Store at 4C.
IgG1 κ
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
MTC754
Unconjugated
10 mM PBS with 0.05% BSA
Mitochondrial fraction of HeLa cells
0.02 mg
Cellular Markers, Mitochondrial Markers